Learn More
Medchemexpress LLC L-histidine, N,1-bis[(1,1-dimethylethoxy)carbonyl]- | 20866-46-0 | MFCD00065577 | >97.0% | 355.39 g/mol | C16H25N3O6 | 25 G

Supplier: Medchemexpress LLC 25GHYW089230
Boc-His(Boc)-OH is the N,N'-di-tert-butoxycarbonyl protected derivative of L-histidine used as an amino acid building block and reagent for peptide and organic synthesis. It is supplied as a white to off-white solid and is commonly used to introduce Boc protection on histidine residues during peptide assembly.
- Boc-protected l-histidine for peptide synthesis.
- Purity >97.0%.
- Molecular weight 355.39 g/mol.
- Chemical formula C16H25N3O6.
- White to off-white solid appearance.
- Available in a 25 G pack; other sizes available upon request.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.