Learn More
Medchemexpress LLC L-Histidine (Standard) | 71-00-1 | 155.15 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN0832R
Encompass_Preferred
L-Histidine (Standard) is the analytical standard of L-Histidine. This product is intended for research and analytical applications. L-Histidine is an essential amino acid for infants and an inhibitor of mitochondrial glutamine transport.
  • Analytical standard grade for reference assays.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Purity of 99.97%.
  • Classified as an amino acid.
  • Endogenous metabolite.
  • Useful as a disease marker for cancer, endocrine diseases, nervous system disorder, cardiovascular system disorder, and immune system disorder.
  • Can be found in microorganisms.
Catalog No. 50-003-89382