Learn More
Medchemexpress LLC L-Histidine (Standard) | 71-00-1 | 155.15 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN0832R50MG
Encompass_Preferred
L-Histidine (Standard) is the analytical standard of L-Histidine, intended for research and analytical applications. It is an essential amino acid for infants and an inhibitor of mitochondrial glutamine transport. This analytical standard is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Intended for research and analytical applications
  • Essential amino acid for infants
  • Inhibitor of mitochondrial glutamine transport
  • Used in qualitative research experiments
  • Used in quantitative research experiments
  • Used in methodological research experiments
  • Applicable in HPLC, GC, and MS
Catalog No. 50-004-15834