Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Histidine (Standard) | 71-00-1 | MFCD00064315 | 155.15 | 100 MG
SDP

Supplier:  Medchemexpress LLC HYN0832R100MG

Encompass_Preferred

L-Histidine (Standard) is the analytical standard of L-Histidine, intended for research and analytical applications. It is an essential amino acid for infants and an inhibitor of mitochondrial glutamine transport.

  • Purity: 99.97%
  • Application: Analytical standard for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Amino acids
  • Initial source: Endogenous metabolite, disease markers, microorganisms.

Catalog No. 50-004-15427


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.