Learn More
Selleck Chemical LLC L-Histidinol dihydrochloride S3337-1g
SDP
Supplier:  Selleck Chemical LLC S33371G
Encompass_Preferred
L-Histidinol dihydrochloride a structural analogue of the essential amino acid L-histidine is a reversible inhibitor of protein biosynthesis which evokes disparate responses from non-tumorigenic and tumorigenic cells in culture
Catalog No. 50-310-9815