Learn More
Cayman Chemical L-HomoargInInhydrochlorIde 1g
SDP
Supplier:  Cayman Chemical 222851 G
Encompass_Preferred
An uncompetitive inhibitor of alkaline phosphatases; inhibits human bone and liver alkaline phosphatases but has no effect on placental or intestinal isoenzymes; inhibits [3H]thymidine uptake by mouse myeloma MOPC 104E cells; inhibits proliferation of C3H/He mouse osteosarcoma cells in vitro; delays tumor growth in a murine C3H/He osteosarcoma model; inhibits high-protein diet-induced pancreatic growth and enzyme secretion in bile-pancreatic juice-diverted rats
Catalog No. 50-298-7898