Learn More
Medchemexpress LLC L-homocysteine, S-[2-[(1-iminoethyl)amino]ethyl]-, phosphate (1:1) | 438542-15-5 | 98.0% | 100 MG

Supplier: Medchemexpress LLC HY12119A100MG
GW274150 phosphate is a potent, selective, orally active NADPH-dependent inhibitor of inducible nitric oxide synthase (iNOS) used in research to probe iNOS-mediated signaling and inflammatory responses (human IC50 = 2.19 μM; Kd = 40 nM).
- Potent iNOS inhibitor with reported human IC50 2.19 μM and Kd 40 nM.
- High reported purity: 98.0% (LCMS).
- Molecular weight 317.30; formula C8H20N3O6PS.
- White to off-white solid; soluble in water and PBS with ultrasonic or heating assistance.
- Storage recommendations: keep dry at 4°C; in solution store -80°C (6 months) or -20°C (1 month).
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.