Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-homocysteine, S-[2-[(1-iminoethyl)amino]ethyl]-, phosphate | 438542-15-5 | 98.0% | 1 ML
SDP

Supplier:  Medchemexpress LLC HY12119A10MM 1ML

Encompass_Preferred

GW274150 phosphate is the phosphate salt form of a selective inducible nitric oxide synthase (iNOS) inhibitor supplied for research use. It is provided as a 10 mM solution (1 mL) or as solid materials, and is accompanied by a datasheet, certificate of analysis, and safety data sheet.

  • White to off-white solid or clear aqueous solution.
  • Purity 98.0%.
  • Molecular weight 317.30 g/mol.
  • Chemical formula C8H20N3O6PS.
  • Available as 10 mM 1 mL solution or in solid sizes (5 mg-100 mg, larger quantities by quote).
  • Solubility: water 100 mg/mL; PBS 12.5 mg/mL (may require ultrasonic and warming).
  • Storage: store solids at 4°C under inert gas; in solvent store at -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human use.

Catalog No. 50-002-31760


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.