Learn More
Medchemexpress LLC L-homocysteine, S-[2-[(1-iminoethyl)amino]ethyl]-, phosphate | 438542-15-5 | 98.0% | 5 MG
SDP
Supplier:  Medchemexpress LLC HY12119A5MG
Encompass_Preferred
GW274150 phosphate is the phosphate salt of a selective inducible nitric oxide synthase (iNOS) inhibitor supplied as a solid for laboratory research. It is provided at high reported purity and intended for research use only.
  • Selective inhibitor of inducible nitric oxide synthase (iNOS).
  • High purity (98.0%).
  • Provided as a solid phosphate salt for stability and handling.
  • Available in small research quantities suitable for assay development.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-32872