Learn More
Medchemexpress LLC L-homocysteine thiolactone hydrochloride | 31828-68-9 | MFCD00012724 | ≥98.0% | 153.63 g/mol | C4H8ClNOS | 1 ML

Supplier: Medchemexpress LLC HY101404A10MM 1ML
L-homocysteine thiolactone hydrochloride is a cyclic thioester derivative of homocysteine used in biochemical and cellular research to investigate protein modification, thiol chemistry, and amino-acid metabolism. It is used to prepare stock solutions and as a reagent in analytical and cell-based assays.
- Used to study post-translational modification of proteins.
- Molecular weight: 153.63 g/mol.
- Soluble in DMSO and water for flexible solvent choice.
- Stable solid form suitable for storage and handling.
- Supplied in small quantities appropriate for stock preparation.
- Provides guidance for in vitro and in vivo formulations.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.