Learn More
Medchemexpress LLC L-Homoserine | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW00229210MM/1ML
Encompass_Preferred
L-Homoserine is a nonessential chiral amino acid that serves as a precursor for L-Threonine and L-Methionine. This versatile compound finds extensive use across pharmaceutical, agricultural, cosmetic, and fragrance industries.
  • Nonessential chiral amino acid
  • Precursor to essential amino acids, L-Threonine and L-Methionine
  • Wide applications in pharmaceutical, agricultural, cosmetic, and fragrance industries
  • Functions as an endogenous metabolite
  • Relevant to inflammation and immunology research
  • Classified as a human gut microbiota metabolite
  • Applicable in studies involving immune system disorder microorganisms, ketones, aldehydes, acids, and disease markers
Catalog No. 50-004-45910