Learn More
Selleck Chemical LLC L-Hydroorotic acid
SDP
Supplier:  Selleck Chemical LLC S611125MG
Encompass_Preferred
L-Dihydroorotic acid (L-dihydroorotate) is an intermediate in the metabolism of Pyrimidine It is a substrate for Dihydroorotate dehydrogenase (mitochondrial)
Catalog No. 50-000-04079