Learn More
Medchemexpress LLC L-Hydroxy arginine d 10mg | 133565-48-7 | 10 MG

Supplier: Medchemexpress LLC HY115411A10MG
L-hydroxy arginine dihydrochloride is an intermediate in the catabolism of L-arginine and a substrate for nitric oxide synthase. The compound is supplied as the dihydrochloride salt in powdered form for research use and is handled and stored under low-temperature, dry conditions.
- CAS number: 133565-48-7.
- Molecular formula: C6H16Cl2N4O3.
- Molecular weight: 263.12 g/mol.
- Physical form: powder.
- Storage: sealed and away from moisture; powder stable at -80°C (2 years) and -20°C (1 year).
- Intended use: for research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.