Learn More
Medchemexpress LLC L-Hydroxy arginine d 25mg | 133565-48-7 | 263.12 g/mol | C6H16Cl2N4O3 | 25 MG

Supplier: Medchemexpress LLC HY115411A25MG
L-hydroxy arginine dihydrochloride is the dihydrochloride salt of Nω-hydroxy-L-arginine, an intermediate in L-arginine catabolism and a substrate for nitric oxide synthase (NOS). Supplied as a solid for research use, it is used in biochemical assays and enzymatic studies of nitric oxide metabolism; follow recommended cold, sealed storage for long-term stability.
- Intermediary in L-arginine catabolism and NOS substrate.
- CAS 133565-48-7.
- Molecular weight 263.12 g/mol.
- Chemical formula C6H16Cl2N4O3.
- High purity (COA shows 99.27%).
- Solid powder form; soluble in water.
- Typical research pack: 25 MG.
- Store sealed, away from moisture; long-term storage at -20°C to -80°C.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.