Learn More
Medchemexpress LLC L-Hydroxy arginine d 50mg | 133565-48-7 | 263.12 | C6H16Cl2N4O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY115411A50MG
Encompass_Preferred
L-Hydroxy arginine dihydrochloride is an intermediate in the catabolism of L-arginine and a substrate for nitric oxide (NO) synthase. Supplied as a white to off-white powder for research use only, the product is provided in a 50 mg package and should be stored sealed and protected from moisture. Typical storage conditions for the powder are -80°C (2 years) or -20°C (1 year).
  • Intermediate in L-arginine catabolism.
  • Substrate for nitric oxide (NO) synthase.
  • Supplied as a powder suitable for in vitro research use.
  • Storage: sealed, away from moisture; powder: -80°C (2 years) or -20°C (1 year).
  • High purity: purity reported as 99.27% by LC-MS for tested batches.
Catalog No. 50-002-28148