Learn More
Medchemexpress LLC L-hydroxyproline-d3 | 1356016-86-8 | 98.7% | 134.15 | C5H6D3NO3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY40135S10MG
Encompass_Preferred
L-hydroxyproline-d3 is the deuterium-labeled form of L-hydroxyproline used as a stable-isotope reference standard and a chiral building block for synthesis and analytical workflows. It is provided in small, exact-quantity packages suitable for method development, isotopic tracing, and synthetic chemistry applications.
  • stable isotope-labeled for accurate mass spectrometry quantitation
  • high purity suitable for analytical use
  • small package sizes minimize waste during method development
  • suitable as a chiral building block in pharmaceutical synthesis
  • delivered with lot-specific certificate of analysis for traceability
Catalog No. 50-002-97944