Learn More
Medchemexpress LLC L-Hyoscyamine | 101-31-5 | 289.38 | 1 ML

Supplier: Medchemexpress LLC HYN047110MM/1ML
L-Hyoscyamine is a naturally occurring plant tropane alkaloid that acts as a potent and competitive muscarinic receptor antagonist. It is the levo-isomer of Atropine. This product is provided as a 10 mM solution in DMSO, ready for use in various research applications. It boasts a high purity of 99.96%, making it ideal for biological research.
- Potent muscarinic receptor antagonist
- Levo-isomer of Atropine
- Purity of 99.96%
- Provided as a 10 mM solution in DMSO
- Suitable for in vitro and in vivo studies
- Stable for 2 years in solvent at -80°C and 1 year at -20°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.