Learn More
Medchemexpress LLC L-Inosine | 21138-24-9 | 99.9% | 268.23 | 1 ML
Supplier: Medchemexpress LLC HY14816610MM/1ML
L-Inosine is the L-configuration of Inosine, an endogenous purine nucleoside produced by catabolism of adenosine. It exhibits a range of biological activities.
- Anti-inflammatory effects
- Antinociceptive effects
- Immunomodulatory effects
- Neuroprotective effects