Learn More
Cayman Chemical L-IsoleucIn-13C6 25mg
SDP
Supplier:  Cayman Chemical 900409025 MG
Encompass_Preferred
An internal standard for the quantification of L-isoleucine by GC- or LC-MS
Catalog No. 50-289-6035