Learn More
Research Products International Corp L-Isoleucine 1 KG
SDP
Supplier:  Research Products International Corp I540201000.0
Encompass_Preferred
L-Isoleucine is crucial for protein synthesis, contributing to the structure and function of proteins within cells. 
It is incorporated into polypeptide chains during translation and is used in cell culture media, supporting the growth and viability of cells. It is particularly important in the cultivation of mammalian cells for the production of recombinant proteins.
L-Isoleucine is commonly utilized in laboratory research, especially in studies related to protein metabolism, cell biology, and molecular biology. Its significance as an essential amino acid makes it a valuable tool in understanding cellular processes.
Catalog No. 50-223-0250