Learn More
Medchemexpress LLC L-isoleucine-13C6,15N,d10 | 2483829-34-9 | 148.18 | 13C6H3D1015NO2 | 10 MG

Supplier: Medchemexpress LLC HYN0771S510MG
L-Isoleucine-13C6,15N,d10 is the deuterium-, 13C-, and 15N-labeled form of the essential amino acid L-isoleucine. It is used as a stable isotope tracer and internal standard to distinguish labeled isoleucine from endogenous sources in analytical workflows such as mass spectrometry and NMR.
- Isotope labeling: 13C6, 15N, and d10.
- Molecular formula: 13C6H3D1015NO2.
- Molecular weight: 148.18.
- CAS number: 2483829-34-9.
- Available pack sizes include 10 mg (other sizes available).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.