Learn More
Medchemexpress LLC L-isoleucine-15N | 59935-30-7 | MFCD00144615 | 98.0% | 132.17 g/mol | C6H13(15N)O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN0771S210MG
Encompass_Preferred
L-Isoleucine-15N is the 15N-labeled form of the essential amino acid L-isoleucine, supplied as a white to off-white solid for research use. It is provided at high purity for applications such as metabolic tracing, isotope labeling studies, and mass spectrometry internal standards.
  • 15N-labeled L-isoleucine for isotope tracing and labeling studies.
  • High purity (98.0%) suitable for analytical applications.
  • Molecular weight 132.17 g/mol for accurate mass calculations.
  • Available in a 10 MG pack size for small-scale laboratory experiments.
  • White to off-white solid, stored sealed away from moisture and light.
  • Data sheet and certificate of analysis available to support quality and batch-specific information.
Catalog No. 50-002-96600