Learn More
Medchemexpress LLC L-Isoleucine-15N,d10 | 142.23 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN0771S7
Encompass_Preferred
L-Isoleucine-15N,d10 is the deuterium and 15N-labeled L-Isoleucine, which is a nonpolar hydrophobic and essential amino acid. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during drug development. Deuteration has garnered attention for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium and 15N-labeled L-Isoleucine
  • Nonpolar hydrophobic amino acid
  • Essential amino acid
  • Used as tracers for quantitation during drug development
  • Influences pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-89385