Learn More
U.S. Pharmacopeia L-Isoleucine, 73-32-5, MFCD00064222, 200mg
Supplier:  U.S. Pharmacopeia 1349502
Encompass
Molecular formula C6H13NO2, Molecular weight 131.17, Sublimes at 168-170 °C
Catalog No. NC0190406