Learn More
Medchemexpress LLC L-Isoleucine-d | 127290-60-2 | 132.18 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN0771S8100MG
Encompass_Preferred
L-Isoleucine-d is a deuterium labeled, nonpolar hydrophobic and essential amino acid. It is suitable for use as a tracer and internal standard in quantitative analysis methods such as NMR, GC-MS, or LC-MS. Deuterium labeling, incorporating stable heavy isotopes, offers the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled
  • Nonpolar hydrophobic amino acid
  • Essential amino acid
  • Functions as a tracer
  • Serves as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-16394