Learn More
Medchemexpress LLC L-Isoleucine (Standard) | 73-32-5 | 131.18 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN0771R100MG
Encompass_Preferred
L-Isoleucine (Standard) is the analytical standard for L-Isoleucine, designed for research and analytical applications. It is an orally active, branched-chain amino acid, specifically the L-enantiomer of isoleucine. This compound functions as a metabolite in various organisms including Saccharomyces cerevisiae, Escherichia coli, plants, humans, algae, and mice. It plays a role in regulating the inflammatory response, offering protection against pathogens both in vivo and in vitro.
  • The compound serves as an analytical standard, providing a reference for assay purposes.
  • It is widely utilized in qualitative, quantitative, and methodological research experiments involving techniques such as HPLC, GC, and MS.
Catalog No. 50-004-15402