Learn More
Medchemexpress LLC L-isoleucinol (2S,3S)-2-amino-3-methylpentan-1-ol | 24629-25-2 | MFCD00004731 | 99.9% | 117.19 g/mol | C6H15NO | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01587510G
Encompass_Preferred
L-Isoleucinol ((2S,3S)-2-amino-3-methylpentan-1-ol) is a chiral amino alcohol used as a biochemical reagent in life-science research. Supplied at high purity, it serves as a building block in organic synthesis, stereochemical investigations, and biochemical assays that require enantiomerically defined amino-alcohols.
  • High purity (99.88%).
  • Defined stereochemistry (2S,3S), suitable for enantioselective studies.
  • Low molecular weight amino alcohol (117.19 g/mol) with formula C6H15NO.
  • Compatible with organic synthesis and assay development workflows.
  • Available in small research quantities for laboratory use.
Catalog No. 50-003-00944