Learn More
Medchemexpress LLC L-Isoleucyl-L-valine | 41017-96-3 | 95.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY12926350MG
Encompass_Preferred
L-Isoleucyl-L-valine (Ile-Val) is a dipeptide known for its role in biological processes. It is recognized for its ability to induce the bovine trypsinogen-trypsin transition.
  • A dipeptide
  • Induces bovine trypsinogen-trypsin transition
  • Appears as a solid, white to off-white
  • Soluble in H2O at 8.33 mg/mL (36.17 mM; requires ultrasonic)
Catalog No. 50-003-41226