Learn More
Medchemexpress LLC L-Isoleucyl-L-valine (Ile-Val) | 41017-96-3 | 95.0% | 25 MG

Supplier: Medchemexpress LLC 25MGHY129263
L-Isoleucyl-L-valine (Ile-Val) is a dipeptide that induces the bovine trypsinogen-trypsin transition. It is for research use only. This product has a molecular weight of 230.30, a formula of C11H22N2O3, and appears as a solid, white to off-white in color.
- Induces bovine trypsinogen-trypsin transition
- Molecular weight: 230.30
- Formula: C11H22N2O3
- Appearance: Solid
- Color: White to off-white
- Ships at room temperature
- Powder storage: -20°C for 3 years or 4°C for 2 years
- In-solvent storage: -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.