Learn More
Medchemexpress LLC L-isoleucyl-L-valine (Ile-Val) | 41017-96-3 | MFCD00038332 | 95.0% | 230.30 g/mol | C11H22N2O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1292635MG
Encompass_Preferred
L-isoleucyl-L-valine (Ile-Val) is a dipeptide composed of L-isoleucine and L-valine that induces the bovine trypsinogen-to-trypsin transition. It is supplied as a solid research reagent (CAS 41017-96-3) with typical purity of 95.0% and a molecular weight of 230.30 g/mol.
Catalog No. 50-002-64060