Learn More
Medchemexpress LLC L-isoleucylglycine | 868-28-0 | MFCD00037777 | ≥95.0% | 188.22 g·mol⁻¹ | C8H16N2O3 | 250 MG

Supplier: Medchemexpress LLC 250MGHYW047902
L-isoleucylglycine is an amino acid dipeptide (isoleucyl-glycine) used as a research reagent and building block in peptide synthesis and biochemical studies. It is supplied as a solid (white to off-white) with typical purity ≥95% and a molecular weight of 188.22 g·mol⁻¹ (CAS 868-28-0). Common pack sizes include small laboratory quantities suitable for analytical and synthetic work.
- Solid, white to off-white physical form
- Purity ≥95%
- Molecular weight 188.22 g·mol⁻¹
- CAS number 868-28-0
- Common pack sizes: 250 mg, 500 mg, 1 g
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.