Learn More
Medchemexpress LLC L-isovaline | 595-40-4 | MFCD00145255 | 99.9% | 117.15 g/mol | C5H11NO2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW018850100MG
Encompass_Preferred
L-Isovaline is a non-proteinogenic amino acid and valine derivative supplied for research use. It is commonly employed in biochemical and pharmacological studies, receptor modulation experiments, and as an analytical reference standard for method development.
  • Non-proteinogenic valine derivative suitable for biochemical research.
  • High purity (99.9%) for analytical and experimental consistency.
  • Solid, white to off-white appearance for easy handling and storage.
  • Well-characterized physicochemical data (MW 117.15 g/mol; CAS 595-40-4).
  • Soluble in water and DMSO with reported guidance for sample preparation.
Catalog No. 50-003-02062