Learn More
Cayman Chemical L-KynurenIn-d4 1mg
SDP
Supplier:  Cayman Chemical 363071 MG
Encompass_Preferred
An internal standard for the quantification of kynurenine by GC- or LC-MS
Catalog No. 50-291-2592