Learn More
Cayman Chemical L-KynurenIn-d4 500ug
SDP
Supplier:  Cayman Chemical 36307500 UG
Encompass_Preferred
An internal standard for the quantification of kynurenine by GC- or LC-MS
Catalog No. 50-291-2383