Learn More
Medchemexpress LLC L-kynurenine | 2922-83-0 | MFCD00069912; MFCD00133224 | 99.9% | 208.21 g/mol | C10H12N2O3 | 1 ML

Supplier: Medchemexpress LLC HY10402610MM 1ML
L-kynurenine is a metabolite of L-tryptophan and an aryl hydrocarbon receptor (AHR) agonist supplied for research applications. It is available as a solid or as a ready-to-use 10 mM solution (1 mL) in DMSO, and is commonly used in metabolic pathway studies, receptor pharmacology, and analytical assays.
- High purity (~99.9%) suitable for research
- Molecular weight 208.21 g/mol
- Chemical formula C10H12N2O3
- Available as solid (50 mg-5 g) or 10 mM 1 mL solution in DMSO
- Solubility: DMSO 20.83 mg/mL (100.04 mM); water 20 mg/mL (96.06 mM)
- Appearance white to yellow solid
- Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.