Learn More
Medchemexpress LLC L-Kynurenine-d4-1 hy 5mg | 5MG
SDP
Supplier:  Medchemexpress LLC HY104026S25MG
Encompass_Preferred
L-Kynurenine-d4-1 hydrochloride is the deuterium labeled L-Kynurenine (HY-104026) L-Kynurenine is a metabolite of the amino acid L-tryptophan L-Kynurenine is an aryl hydrocarbon receptor agonist[1 [2
Catalog No. 50-002-42552