Learn More
Cayman Chemical L L-DItyrosIn 10mg
SDP
Supplier:  Cayman Chemical 3574610 MG
Encompass_Preferred
An amino acid conjugate; cross-links of it can form between fibrinogen molecules in the presence of hematin and plasma-generated hydrogen peroxide under non-thermal plasma exposure; cross-links of it can form between human amylin peptides in the presence of copper (II) ions and hydrogen peroxide
Catalog No. 50-291-0212