Learn More
Sigma Aldrich Fine Chemicals Biosciences L-Lactate Dehydrogenase L-
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 10127230001
Encompass
L-lactateNAD oxidoreductase.L-lactate dehydrogenase catalyzes the reversible reduction of pyruvate to L-lactate.
Catalog No. 50-290-1122