Learn More
Sigma Aldrich Fine Chemicals Biosciences L Lactic Dehydrogenase fro
Supplier:  Sigma Aldrich Fine Chemicals Biosciences L1006
Encompass
L-Lactic Dehydrogenase (LDH) is present in the cell cytoplasm and is part of the glycolytic pathway. It exists in different isoforms in heart and muscles. LDH is a tetramer corresponding to a molecular weight of 140 kDa. LDH has a nucleotide-cofactor binding site and has the structure of Rossmann fold.
Catalog No. 50-290-8475