Learn More
Cayman Chemical L-Leu-AMChydrochlorIde 500mg
SDP
Supplier:  Cayman Chemical 34267500 MG
Encompass_Preferred
A fluorogenic substrate for leucine aminopeptidase; AMC is released upon enzymatic cleavage by leucine aminopeptidase and its fluorescence can be used to quantify leucine aminopeptidase activity; ex/em = 340-360/440-460 nm, respectively
Catalog No. 50-295-0181