Learn More
Cayman Chemical L-LeucIn-d10 5mg
SDP
Supplier:  Cayman Chemical 348435 MG
Encompass_Preferred
An internal standard for the quantification of L-leucine by GC- or LC-MS
Catalog No. 50-291-2655