Learn More
Medchemexpress LLC L-Leucine-13C | 74292-94-7 | 132.17 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN0486S1100MG
Encompass_Preferred
L-Leucine-13C is the 13C-labeled L-Leucine, an essential branched-chain amino acid (BCAA) that activates the mTOR signaling pathway. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • 13C-labeled L-Leucine
  • Activates the mTOR signaling pathway
  • Used as a tracer
  • Used as an internal standard for quantitative analysis
  • Essential branched-chain amino acid (BCAA)
Catalog No. 50-003-89875