Learn More
Medchemexpress LLC L-Leucine-18O2 | 73579-45-0 | 135.17 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN0486S10100MG
Encompass_Preferred
L-Leucine-18O2 is an 18O-labeled L-Leucine, an essential branched-chain amino acid (BCAA) known for activating the mTOR signaling pathway. This compound is suitable for various applications, including use as a tracer and as an internal standard.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Isotope-labeled compound
  • Activates the mTOR signaling pathway
  • Essential branched-chain amino acid
Catalog No. 50-003-89076