Learn More
Medchemexpress LLC L-Leucine-18O2 | 73579-45-0 | 135.17 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN0486S1050MG
Encompass_Preferred
L-Leucine-18O2 is an 18O-labeled L-Leucine, an essential branched-chain amino acid (BCAA) that activates the mTOR signaling pathway. It is intended for research use only and not for sale to patients. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-89081