Learn More
Selleck Chemical LLC L-Leucine S3753-1g
SDP
Supplier:  Selleck Chemical LLC S37531G
Encompass_Preferred
Leucine ((S)-Leucine Leu) is one of nine essential amino acids in humans which is important for protein synthesis and many metabolic functions It contributes to regulation of blood-sugar levels growth and repair of muscle and bone tissue growth hormone production and wound healing L-Leucine is an essential branched-chain amino acid (BCAA) which activates the mTOR signaling pathway
Catalog No. 50-310-9285