Learn More
Medchemexpress LLC L-Leucine-7-amido-4-methylcoumarin hydrochloride | 62480-44-8 | 99.98% | 324.80 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY137844
Encompass_Preferred
L-Leucine-7-amido-4-methylcoumarin (Leu-AMC) hydrochloride is a bright blue fluorogenic peptidyl substrate for LAP3 (leucine aminopeptidase). It is utilized for leucine aminopeptidase inhibition assays in vitro.
  • Bright blue fluorogenic peptidyl substrate
  • Utilized for LAP3 (leucine aminopeptidase)
  • Can be used for leucine aminopeptidase inhibition assays in vitro
  • Hydrolyzed 7-amino-4-methylcoumarin (AMC) can be measured using a fluorescence spectrophotometer (Ex/Em=365/440 nm)
Catalog No. 50-003-49788