Learn More
Medchemexpress LLC L-Leucine-d10 | 106972-44-5 | 141.23 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN0486S5MG
Encompass_Preferred
L-Leucine-d10 is a deuterium-labeled L-Leucine, an essential branched-chain amino acid (BCAA) that activates the mTOR signaling pathway. This compound is intended for research use only and can serve as a tracer or an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS. Deuteration, which incorporates stable heavy isotopes, is recognized for its potential to influence the pharmacokinetic and metabolic profiles of drugs, making L-Leucine-d10 valuable in drug development processes.
  • Deuterium labeled L-Leucine
  • Essential branched-chain amino acid
  • Activates the mTOR signaling pathway
  • For research use only
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-90129