Learn More
Medchemexpress LLC L-leucine, glycyl- | 869-19-2 | MFCD00008127 | 99.5% | 188.22 | C8H16N2O3 | 100 G

Supplier: Medchemexpress LLC HYW016077100G
Glycyl-L-leucine is a dipeptide of glycine and L-leucine used as a biochemical substrate and research standard. It is supplied as a characterized solid with documented identity, purity, solubility, and storage recommendations for laboratory applications.
- Molecular formula C8H16N2O3 and molecular weight 188.22.
- High purity >99.4% (99.49% by HPLC).
- Soluble in water 25 mg/mL (132.82 mM); PBS 50 mg/mL (265.65 mM) with ultrasonic or warming.
- Recommended storage: powder -20°C (3 years) or 4°C (2 years); solutions -80°C (6 months), -20°C (1 month).
- Prepared stock solution guidance provided for common concentrations (1-10 mM).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.