Learn More
Medchemexpress LLC L-Leucine, N-[4-[(1H-imidazol-5-ylmethyl)amino]-2-(1-naphthalenyl)benzoyl]- | 191102-79-1 | 98.5% | 456.54 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY12369
Encompass_Preferred
GGTI-2133 is a peptidomimetic and a selective geranylgeranyltransferase I (GGTase-I) inhibitor with an IC50 of 38 nM. It exhibits 140-fold selectivity over farnesyltransferase (FTase) (IC50 of 5.4 μM). GGTI-2133 inhibits the geranylation of Rap-1A with an IC50 of 10 μM. It can be utilized for the study of eosinophilic airway inflammation, such as asthma.
  • Selective geranylgeranyltransferase I (GGTase-I) inhibitor.
  • Shows 140-fold selectivity over farnesyltransferase (FTase).
  • Inhibits the geranylation of Rap-1A.
  • Can be used for the study of eosinophilic airway inflammation such as asthma.
  • Inhibits infiltration of inflammatory cells into airways in mouse experimental asthma (at 5 mg/kg; intraperitoneal injection; once a day; for 10 days).
Catalog No. 50-004-38326