Learn More
Medchemexpress LLC L-leucine, N-[(9H-fluoren-9-ylmethoxy)carbonyl]-2-methyl | 312624-65-0 | 99.5% | 367.44 g/mol | C22H25NO4 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0487395G
Encompass_Preferred
Fmoc-α-Me-Leu-OH is an Fmoc-protected, alpha-methylated leucine derivative used as a protected amino acid building block for solid-phase peptide synthesis and peptide research. It is supplied as a solid powder, with analytical documentation available in the manufacturer COA and datasheet.
  • Fmoc-protected amino acid for solid-phase peptide synthesis.
  • Alpha-methyl substitution introduces conformational constraint in peptides.
  • High purity: 99.5% (as supplied).
  • Molecular formula C22H25NO4; molecular weight 367.44 g/mol.
  • Supplied as a dry solid in small gram-scale packages for research use.
Catalog No. 50-002-10113